Skip to Content
Merck
All Photos(3)

Key Documents

246093

Sigma-Aldrich

Methyl butyrate

99%

Synonym(s):

3-Methylpropanoic acid methyl ester, Methyl butanoate, Methyl n-butyrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$71.90
100 ML
$76.60
500 ML
$96.60

$71.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$71.90
100 ML
$76.60
500 ML
$96.60

About This Item

Linear Formula:
CH3CH2CH2COOCH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740743
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

3.5 (vs air)

Quality Level

vapor pressure

40 mmHg ( 30 °C)

Assay

99%

form

liquid

expl. lim.

3.5 %

refractive index

n20/D 1.385 (lit.)

bp

102-103 °C (lit.)

solubility

water: soluble 60 part
alcohol: miscible
diethyl ether: soluble

density

0.898 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCC(=O)OC

InChI

1S/C5H10O2/c1-3-4-5(6)7-2/h3-4H2,1-2H3

InChI key

UUIQMZJEGPQKFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl butyrate, one of the odorous compound present in different types of adhesive (hotmelt, vinyl acetate ethylene, starch, polyvinyl acetate and acrylic) used in food packaging, was identified by GC-O-MS (gas chromatography–mass spectrometry–olfactometry)[1].

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

53.0 °F

Flash Point(C)

11.67 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Twiner et al.
Genomics, 91(3), 289-300 (2008-01-15)
Azaspiracid-1 (AZA-1) is a marine biotoxin reported to accumulate in shellfish from several countries, including eastern Canada, Morocco, and much of western Europe, and is frequently associated with severe gastrointestinal human intoxication. As the mechanism of action of AZA-1 is
Kaixin Li et al.
Physical chemistry chemical physics : PCCP, 18(48), 32723-32734 (2016-11-24)
Brønsted acidic ionic liquids (BAILs) can play a dual role, as a solvent and as a catalyst, in many reactions. However, molecular details of the catalytic mechanism are poorly understood. We present here a density functional theory (DFT) study for
Sang-Ryong Lee et al.
The Science of the total environment, 610-611, 992-996 (2017-08-26)
This report proposes a new approach to evaluate the odour nuisance of cattle manure samples from three different cattle breeds (i.e., native cattle, beef cattle, and milk cow) by means of quantification and speciation of volatile fatty acids (VFAs). To
S H Sterri et al.
Biochemical pharmacology, 34(15), 2779-2785 (1985-08-01)
A radiochemical assay for carboxylesterase based on the substrate methyl[1-14C]butyrate is described. The blank value corresponds to 1.04 micrograms (liver)-1.44 mg (plasma) of tissues with the highest and lowest activity respectively, which constitute the sensitivity of the method. The hydrolysis
Zheng Wu et al.
Neuron, 106(2), 316-328 (2020-02-28)
Cognitive capacities afford contingent associations between sensory information and behavioral responses. We studied this problem using an olfactory delayed match to sample task whereby a sample odor specifies the association between a subsequent test odor and rewarding action. Multi-neuron recordings

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service