Skip to Content
Merck
All Photos(3)

Key Documents

246085

Sigma-Aldrich

Ethyl isobutyrate

99%

Synonym(s):

2-Methylpropanoic acid ethyl ester, 2-Methylpropionic acid ethyl ester, Ethyl 2,2-dimethylacetate, Ethyl 2-methyl-1-propanoate, Ethyl 2-methylpropanoate, Ethyl 2-methylpropionate, Ethyl isobutanoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$36.60
500 ML
$83.70

$36.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$36.60
500 ML
$83.70

About This Item

Linear Formula:
(CH3)2CHCOOC2H5
CAS Number:
Molecular Weight:
116.16
Beilstein:
773846
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$36.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

4.01 (vs air)

vapor pressure

40 mmHg ( 33.8 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.387 (lit.)

bp

112-113 °C (lit.)

solubility

alcohol: miscible(lit.)
diethyl ether: miscible(lit.)
water: slightly soluble(lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(C)C

InChI

1S/C6H12O2/c1-4-8-6(7)5(2)3/h5H,4H2,1-3H3

InChI key

WDAXFOBOLVPGLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl isobutyrate is one of the odorants found in Tuber indicum[1]. It exhibits high flavor dilution factor (FD) value[2].

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Culleré et al.
Food chemistry, 141(1), 105-110 (2013-06-19)
The Tuber indicum (Chinese truffle) and Tuber melanosporum (Black truffle) species are morphologically very similar but their aromas are very different. The black truffle aroma is much more intense and complex, and it is consequently appreciated more gastronomically. This work
Sonia Collin et al.
Journal of agricultural and food chemistry, 60(1), 380-387 (2011-11-29)
The aromatic profile of Jura flor-sherry wines (also called "yellow wines") has been little studied. Only acetaldehyde, diethoxy-1,1-ethane, and sotolon have been described as key odorants. In the present work, three wines (vintages 2002 and 2003) were investigated by gas
Gabriella Lo Feudo et al.
Journal of agricultural and food chemistry, 58(6), 3801-3807 (2010-02-23)
The concentration of 32 elements (Al, As, Ba, Be, Ca, Cd, Ce, Cu, Dy, Fe, K, La, Lu, Mg, Mn, Na, Nd, Pb, Rb, Sm, Sr, Th, U, V, Zn) was determined in tomatoes harvested in different four Italian regions
Ting Zhou et al.
Food chemistry, 335, 127664-127664 (2020-08-03)
Odor-taste interaction has become a popular salt reduction method. In this study, the odorants associated with saltiness in soy sauce were selected by gas chromatography/olfactometry-associated taste (GC/O-AT), and their ability to induce saltiness/umami enhancement was verified by sensory evaluation. A
Tanya E Rutan et al.
Journal of agricultural and food chemistry, 66(38), 10053-10066 (2018-09-04)
The effects of two cluster thinning regimes (low and moderate) with Vitis vinifera cv. Pinot noir, in vineyards located in Central Otago, New Zealand, on wine composition, were studied across three consecutive seasons. There were strong correlations between the extent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service