Skip to Content
Merck
All Photos(1)

Key Documents

291137

Sigma-Aldrich

2-Amino-1-naphthalenesulfonic acid

98%

Synonym(s):

2-Naphthylamine-1-sulfonic acid, Tobias acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6SO3H
CAS Number:
Molecular Weight:
223.25
Beilstein:
613084
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

SMILES string

Nc1ccc2ccccc2c1S(O)(=O)=O

InChI

1S/C10H9NO3S/c11-9-6-5-7-3-1-2-4-8(7)10(9)15(12,13)14/h1-6H,11H2,(H,12,13,14)

InChI key

GWIAAIUASRVOIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-1-naphthalenesulfonic acid is a manufacturing intermediate in the monosulfo monoazo color additive D&C Red No. 34 and its lakes[1].

Application

2-Amino-1-naphthalenesulfonic acid has been used in the preparation of 2-(2-naphthylamino)benzoxazole by reacting with 2-chlorobenzoxazole[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kumar et al.
British journal of industrial medicine, 38(2), 167-169 (1981-05-01)
Immunological studies have shown an increased lymphocyte reactivity in patients with early stage bladder cancer and individuals with pre-stage T1 exposed to bladder carcinogens (2-naphthylamine and industrial 1-naphthylamine containing 4-8% 2-naphthylamine) before 1952-that is, those at high risk of developing
Bhakti Petigara Harp et al.
Journal of AOAC International, 92(6), 1639-1643 (2010-02-20)
A reversed-phase LC method was developed to determine two manufacturing intermediates in the monosulfo monoazo color additive D&C Red No. 34 and its lakes. The analytes are 2-amino-1-naphthalenesulfonic acid (Tobias acid) and 3-hydroxy-2-naphthalenecarboxylic acid (3-hydroxy-2-naphthoic acid). This method can be
Ayman Ayoub Abdel-Shafi
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(4-5), 1228-1236 (2006-09-05)
The photophysical characteristics of the ground and excited states of 2-naphthylamine-6-sulfonate (2-NA-6-S) were investigated in different solvents and in beta-cyclodextrin (beta-CD). The spectral shifts are well correlated with Kamlet-Taft relationship. Multiple linear regression analysis indicated that both non-specific dipolar interaction
Non-carcinogenicity in mice of a sulfonic acid derivative of 2-naphthylamine.
G Della Porta et al.
Carcinogenesis, 3(6), 647-649 (1982-01-01)
Jolanta Polak et al.
International journal of molecular sciences, 21(6) (2020-03-21)
Novel sustainable processes involving oxidative enzymatic catalysts are considered as an alternative for classical organic chemistry. The unique physicochemical and bioactive properties of novel bio-products can be obtained using fungal laccase as catalyst. Among them are textile biodyes synthesised during

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service