Skip to Content
Merck
All Photos(2)

Key Documents

287970

Sigma-Aldrich

trans-1,2-Bis(diphenylphosphino)ethylene

97%

Synonym(s):

(E)-Vinylenebis[diphenylphosphine], trans-1,2-Ethenediylbis(diphenylphosphine), NSC 132587

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2PCH=CHP(C6H5)2
CAS Number:
Molecular Weight:
396.40
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

126-130 °C (lit.)

functional group

phosphine

SMILES string

c1ccc(cc1)P(\C=C\P(c2ccccc2)c3ccccc3)c4ccccc4

InChI

1S/C26H22P2/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26/h1-22H/b22-21+

InChI key

NCKJIJSEWKIXAT-QURGRASLSA-N

Application

Catalyst for:
  • Coupling of primary and secondary alkyl halides with aryl grignard reagents
  • Homogeneous hydrogenation of Carbon Dioxide mediated by Ruthenium

Reactant for:
  • Preparation of cyclopalladated derivatives of benzylidene
  • Preparation of cyclopalladated complexes containing tridentate thiosemicarbazone ligands as antimalarial agents
  • Catalytic asymetric intramolecular hydroamination of alkynylamines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noora Svahn et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(55), 14654-14667 (2018-08-01)
A series of 4-ethynylaniline gold(I) complexes containing monophosphane (1,3,5-triaza-7-phosphaadamantane (pta; 2), 3,7-diacetyl-1,3,7-triaza-5-phosphabicyclo[3.3.1]nonane (3), and PR3 , with R=naphthyl (4), phenyl (5), and ethyl (6)) and diphosphane (bis(diphenylphosphino)acetylene (dppa; 7), trans-1,2-bis(diphenylphosphino)ethene (dppet; 8), 1,2-bis(diphenylphosphino)ethane (dppe; 9), and 1,3-bis(diphenylphosphino)propane (dppp; 10)) ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service