Skip to Content
Merck
All Photos(1)

Documents

261920

Sigma-Aldrich

1,2-Bis(dichlorophosphino)ethane

97%

Synonym(s):

1,2-Ethanediylbis[phosphonous dichloride], Ethylenebis(dichlorophosphine), P,P′-(1,2-Ethanediyl)bis(dichlorophosphane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2PCH2CH2PCl2
CAS Number:
Molecular Weight:
231.81
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.588 (lit.)

bp

70 °C/1 mmHg (lit.)

density

1.536 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

ClP(Cl)CCP(Cl)Cl

InChI

1S/C2H4Cl4P2/c3-7(4)1-2-8(5)6/h1-2H2

InChI key

SBWAJHLQMFBNIN-UHFFFAOYSA-N

Related Categories

General description

1,2-Bis(dichlorophosphino)ethane is an organophosphorus compound commonly used as a precursor to synthesize ethylene-bridged bis(phosphane)s ligands by reacting with an organometallic reagent.

Application

1,2-Bis(dichlorophosphino)ethane can be used as a precursor to synthesize ethylenebis(phosphonic dichloride).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron-Poor Olefin Polymerization Catalysts Based on Semi-Fluorinated Bis (phosphane) s
European Journal of Inorganic Chemistry, 2000(9), 2063-2070 (2000)
A convenient synthesis of 1, 2-bis (dichlorophosphino) ethane, 1, 2-bis (dimethylphosphino) ethane and 1, 2-bis (diethylphosphino) ethane
Burt RJ, et al.
Journal of Organometallic Chemistry, 182(2), 203-206 (1979)
Cofactor-type inhibitors of inosine monophosphate dehydrogenase via modular approach: targeting the pyrophosphate binding sub-domain
Felczak K, et al.
Bioorganic & Medicinal Chemistry, 9(5), 1594-1605 (2011)
Ruiyan Sun et al.
ChemSusChem, 12(14), 3278-3285 (2019-04-30)
Methyl formate was produced in one pot through the hydrogenation of CO2 to formic acid/formate followed by an esterification step. The route offers the possibility to integrate renewable energy into the fossil-based chemical value chain. In this work, a phosphine-polymer-anchored

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service