Skip to Content
Merck
All Photos(1)

Key Documents

277452

Sigma-Aldrich

Methyl 2-methylbutyrate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$107.00

$107.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$107.00

About This Item

Linear Formula:
CH3CH2CH(CH3)COOCH3
CAS Number:
Molecular Weight:
116.16
Beilstein:
1720409
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$107.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.393 (lit.)

bp

115 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCC(C)C(=O)OC

InChI

1S/C6H12O2/c1-4-5(2)6(7)8-3/h5H,4H2,1-3H3

InChI key

OCWLYWIFNDCWRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 2-methylbutyrate is the main component of the fragrance of Victoria[1]. It is the volatile constituent of osmeterial secretions of 5th larval instars of a swallowtail, Papilio protenor demetrius[2].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential pollinator-attracting chemicals fromVictoria (Nymphaeaceae).
Kite G, et al.
Biochemical Systematics and Ecology, 19(7), 535-539 (1991)
Volatile constituents of larval osmeterial secretions in Papilio protenor demetrius.
Honda K.
Journal of Insect Physiology, 26(1), 39-45 (1980)
Rachel S Leisso et al.
Physiologia plantarum, 153(2), 204-220 (2014-06-20)
'Soggy breakdown' (SB) is an internal flesh disorder of 'Honeycrisp' apple (Malus × domestica Borkh.) fruit that occurs during low temperature storage. The disorder is a chilling injury (CI) in which visible symptoms typically appear after several weeks of storage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service