Skip to Content
Merck
All Photos(1)

Documents

T71803

Sigma-Aldrich

Pivalic acid

99%

Synonym(s):

2,2-Dimethylpropionic acid, Trimethylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCO2H
CAS Number:
Molecular Weight:
102.13
Beilstein:
969480
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.6 (vs air)

vapor pressure

9.75 mmHg ( 60 °C)

Assay

99%

reaction suitability

reaction type: C-H Activation

bp

163-164 °C (lit.)

mp

32-35 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(C(C)(C)C)=O

InChI

1S/C5H10O2/c1-5(2,3)4(6)7/h1-3H3,(H,6,7)

InChI key

IUGYQRQAERSCNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Pivalic acid is a carboxylic acid, employed as a ligand to synthesize cerium(IV) hexanuclear clusters.

Application

Pivalic acid can be employed:
  • As a co-catalyst with palladium for the arylation of unactivated arenes and N-heterocycles.
  • As an additive to facilitate the carbonylative suzuki reactions to synthesize biaryl ketones from aryl iodides and arylboronic acids by using palladium nanoparticles as catalyst.
  • In the cyclization reaction of benzamides with alkynes to synthesize isoquinolones in the presence of 8-aminoquinoline ligand and cobalt catalyst.

Caution

Pungent odour/Stench

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Phosphine-Free, Palladium-Catalyzed Arylation of Heterocycles through C?H Bond Activation with Pivalic Acid as a Cocatalyst.
Zhao D, et al.
Chemistry?A European Journal , 15(6), 1337-1340 (2009)
Asit K Chakraborti et al.
The Journal of organic chemistry, 71(15), 5785-5788 (2006-07-15)
Magnesium bistrifluoromethanesulfonimide catalyzed the acetylation of phenols, alcohols, and thiols under solvent-free conditions at room temperature and in short times. Electron-deficient and sterically hindered phenols provided excellent yields. The catalyst was found to be general for acylation with other anhydrides
Mamoru Tobisu et al.
Chemical communications (Cambridge, England), 47(10), 2946-2948 (2011-01-26)
A nickel-catalyzed reductive deoxygenation of aryl alkyl ethers and aryl pivalates has been developed. Hydrosilanes serve as a mild reducing agent. The present protocol allows the use of a pivalate group as a robust and traceless steering group in arene
Cross-coupling of aryl/alkenyl pivalates with organozinc reagents through nickel-catalyzed C-O bond activation under mild reaction conditions.
Bi-Jie Li et al.
Angewandte Chemie (International ed. in English), 47(52), 10124-10127 (2008-11-19)
Vishal Kandathil et al.
Carbohydrate polymers, 223, 115060-115060 (2019-08-21)
The core characteristics of a perfect catalyst include good activity, simple design, excellent stability, easy recovery from reaction mixture, recyclability, and have the provision for easy scale up. The ease in synthesis, recyclability and scale up makes the dip catalyst

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service