Skip to Content
Merck
All Photos(1)

Key Documents

258598

Sigma-Aldrich

4-Ethoxyphenol

99%

Synonym(s):

Hydroquinone monoethyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$62.20
25 G
$104.00

$62.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$62.20
25 G
$104.00

About This Item

Linear Formula:
C2H5OC6H4OH
CAS Number:
Molecular Weight:
138.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

121 °C/9 mmHg (lit.)

mp

64-67 °C (lit.)

SMILES string

CCOc1ccc(O)cc1

InChI

1S/C8H10O2/c1-2-10-8-5-3-7(9)4-6-8/h3-6,9H,2H2,1H3

InChI key

LKVFCSWBKOVHAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Ethoxyphenol is the major dehalogenated product formed during H2O2-driven microperoxidase-8-catalyzed dehalogenation of 4-fluorophenol[1].

Application

4-Ethoxyphenol has been used as substrate to evaluate kinetic constant for the monophenolase activity of mushroom tyrosinase[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Osman et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(9), 4295-4299 (1997-04-29)
The results of this study report the H2O2-driven microperoxidase-8 (MP8)-catalyzed dehalogenation of halophenols such as 4-fluorophenol, 4-chlorophenol, 4-bromophenol, and 2-fluorophenol in alcoholic solvents. In methanol, the conversion of the para-halophenols and 2-fluorophenol to, respectively, 4-methoxyphenol and 2-methoxyphenol, as the major
J C Espín et al.
European journal of biochemistry, 267(5), 1270-1279 (2000-02-26)
This paper reports a quantitative study of the effect of ring substituents in the 1-position of the aromatic ring on the rate of monophenol hydroxylation and o-diphenol oxidation catalyzed by tyrosinase. A possible correlation between the electron density of the
Cynthia D Selassie et al.
Journal of medicinal chemistry, 48(23), 7234-7242 (2005-11-11)
In this comprehensive study on the caspase-mediated apoptosis-inducing effect of 51 substituted phenols in a murine leukemia cell line (L1210), we determined the concentrations needed to induce caspase activity by 50% (I50) and utilized these data to develop the following
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service