Skip to Content
Merck
All Photos(1)

Key Documents

253928

Sigma-Aldrich

1-Bromo-4-tert-butylbenzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$79.00
100 G
$167.00

$79.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$79.00
100 G
$167.00

About This Item

Linear Formula:
(CH3)3CC6H4Br
CAS Number:
Molecular Weight:
213.11
Beilstein:
1859117
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$79.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

80-81 °C/2 mmHg (lit.)

mp

13-16 °C (lit.)

density

1.229 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CC(C)(C)c1ccc(Br)cc1

InChI

1S/C10H13Br/c1-10(2,3)8-4-6-9(11)7-5-8/h4-7H,1-3H3

InChI key

XHCAGOVGSDHHNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-4-tert-butylbenzene undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents[1].

Application

1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid[2], 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7)[3].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F

Flash Point(C)

97 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel non-symmetric nickel-diimine complexes for the homopolymerization of ethene: Control of branching by catalyst design.
Schmid M, et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 57(10), 1141-1146 (2002)
William F Bailey et al.
The Journal of organic chemistry, 71(7), 2825-2828 (2006-03-25)
The outcome of reactions of 1-bromo-4-tert-butylbenzene (1), a representative aryl bromide, with n-BuLi or t-BuLi at 0 degrees C in a variety of solvent systems has been investigated. The products of reactions of 1 with n-BuLi vary significantly with changes
John W Huffman et al.
Bioorganic & medicinal chemistry, 16(1), 322-335 (2007-10-09)
A series of 1-deoxy analogs of CP-47,497 (8 and 13, n=0-7) and 1-deoxy analogs of CP-55,940 (9, n=0-7) have been synthesized and their affinities for the cannabinoid CB(1) and CB(2) receptors have been determined. Although the majority of these compounds
Xiao Yan et al.
Physical chemistry chemical physics : PCCP, 22(11), 6222-6230 (2020-03-05)
Unveiling the reaction mechanism is significant for developing high-performance catalysts. In this paper, a series of precisely controlled PdxM147-x (M = Cu, Pt, Au, Rh, Ru) dendrimer encapsulated nanoparticles (DENs) has been successfully synthesized. The mechanisms of PdxM147-x as catalysts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service