Skip to Content
Merck
All Photos(2)

Key Documents

480053

Sigma-Aldrich

4-tert-Butylphenylboronic acid

≥95.0%

Synonym(s):

(p-tert-Butylphenyl)boronic acid, 4-t-Butylbenzeneboronic acid, 4-t-Butylphenylboronic acid, 4-tert-Butylbenzeneboronic acid, p-tert-Butylbenzeneboronic acid, [4-(1,1-Dimethylethyl)phenyl]boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4B(OH)2
CAS Number:
Molecular Weight:
178.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95.0%

mp

191-196 °C (lit.)

SMILES string

CC(C)(C)c1ccc(cc1)B(O)O

InChI

1S/C10H15BO2/c1-10(2,3)8-4-6-9(7-5-8)11(12)13/h4-7,12-13H,1-3H3

InChI key

MNJYZNVROSZZQC-UHFFFAOYSA-N

Application

Cross-coupling building block used in synthesis of tetracycline derivatives.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 68, 10199-10199 (2003)
Guillaume Erbland et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(71), 16328-16339 (2019-10-12)
The design and synthesis of two families of molecular-gear prototypes is reported, with the aim of assembling them into trains of gears on a surface and ultimately achieving controlled intermolecular gearing motion. These piano-stool ruthenium complexes incorporate a hydrotris(indazolyl)borate moiety
Xiao Yan et al.
Physical chemistry chemical physics : PCCP, 22(11), 6222-6230 (2020-03-05)
Unveiling the reaction mechanism is significant for developing high-performance catalysts. In this paper, a series of precisely controlled PdxM147-x (M = Cu, Pt, Au, Rh, Ru) dendrimer encapsulated nanoparticles (DENs) has been successfully synthesized. The mechanisms of PdxM147-x as catalysts
Heng Wang et al.
Chirality, 27(8), 523-531 (2015-04-25)
A novel pyridineoxazoline (PyOx) containing helical polymer, poly{(-)-(S)-4-tert-butyl-2-[5-(4-tert-butylphenyl)-3-vinylpyridin-2-yl]-oxazoline} (PA), was designed and synthesized to approach the effect of chain conformation on the catalytic property. Its complex with Cu(OTf)(2) , i.e., Cu(II)-PA, was employed to catalyze the homogeneous Diels-Alder (D-A) reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service