Skip to Content
Merck
All Photos(1)

Key Documents

290157

Sigma-Aldrich

1,3-Bis(trifluoromethyl)-5-bromobenzene

97%, contains 3% Copper Shot (CAS 7440-50-8) as stabilizer

Synonym(s):

1-Bromo-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)bromobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$81.80
50 G
$202.00

$81.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$81.80
50 G
$202.00

About This Item

Linear Formula:
(CF3)2C6H3Br
CAS Number:
Molecular Weight:
293.00
Beilstein:
2123669
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$81.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

contains

3% Copper Shot (CAS 7440-50-8) as stabilizer

refractive index

n20/D 1.427 (lit.)

bp

154 °C (lit.)

mp

−16 °C (lit.)

density

1.699 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1cc(Br)cc(c1)C(F)(F)F

InChI

1S/C8H3BrF6/c9-6-2-4(7(10,11)12)1-5(3-6)8(13,14)15/h1-3H

InChI key

CSVCVIHEBDJTCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Bis(trifluoromethyl)-5-bromobenzene was used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion which is a stabilizing counterion for highly electrophilic organic[1] and organometallic cations[1][2].

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilfried Posch et al.
The Journal of allergy and clinical immunology, 147(6), 2083-2097 (2021-04-15)
Excessive inflammation triggered by a hitherto undescribed mechanism is a hallmark of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infections and is associated with enhanced pathogenicity and mortality. Complement hyperactivation promotes lung injury and was observed in patients suffering from
The Journal of Organic Chemistry, 57, 5545-5545 (1992)
Organometallics, 11, 3920-3920 (1992)
Sarah Davidson et al.
Cell reports, 31(7), 107628-107628 (2020-05-21)
Here, using single-cell RNA sequencing, we examine the stromal compartment in murine melanoma and draining lymph nodes (LNs) at points across tumor development, providing data at http://www.teichlab.org/data/. Naive lymphocytes from LNs undergo activation and clonal expansion within the tumor, before

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service