Skip to Content
Merck
All Photos(1)

Key Documents

251941

Sigma-Aldrich

2-Chlorophenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NCO
CAS Number:
Molecular Weight:
153.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.5575 (lit.)

bp

83-84 °C/10 mmHg (lit.)

solubility

water: insoluble

density

1.273 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1N=C=O

InChI

1S/C7H4ClNO/c8-6-3-1-2-4-7(6)9-5-10/h1-4H

InChI key

NOHQUGRVHSJYMR-UHFFFAOYSA-N

Application

2-Chlorophenyl isocyanate has been used:
  • as standard to investigate the photocatalytic degradation of Monuron over TiO2 suspensions
  • in the preparation of 2-(2-chlorophenylamino)-3,5-dihydro-5,5-dimethyl-3-phenyl-6H-imidazo[1,2-b]-1,2,4-triazol-6-one
  • in synthesis of pentenylated β-cyclodextrin chlorophenyl carbamates

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Facile Synthesis of 3, 5-Dihydro-6H-imidazo [1, 2-b]-1, 2, 4-triazol-6-ones by an Iminophosphorane-Mediated Annulation.
Yuan J-Z, et al.
European Journal of Organic Chemistry, 2006(18), 4170-4176 (2006)
Photocatalytic degradation of monuron in aqueous titanium dioxide dispersions.
Pramauro E, et al.
Environmental Science & Technology, 27(9), 1790-1795 (1993)
Monochloro-substituted phenyl carbamoylated ?-cyclodextrins as p-acid chiral stationary phases for high-performance liquid chromatography.
Bai ZW, et al.
Chromatographia, 58(1-2), 43-46 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service