Skip to Content
Merck
All Photos(1)

Documents

227269

Sigma-Aldrich

Benzyl isocyanate

99%

Synonym(s):

α-Isocyanatotoluene, (Isocyanatomethyl)benzene, 1-(Isocyanatomethyl)benzene, Phenylmethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NCO
CAS Number:
Molecular Weight:
133.15
Beilstein:
636508
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

101-104 °C/33 mmHg (lit.)

density

1.078 g/mL at 25 °C (lit.)

SMILES string

O=C=NCc1ccccc1

InChI

1S/C8H7NO/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

YDNLNVZZTACNJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl isocyanate inhibits the formation of benzo[a]pyrene (BP)-DNA adduct and posses chemopreventive properties.

Application

Benzyl isocyanate was used to investigate the kinetics of isocyanate derivatization reaction in a continuous flow glass microfluidics chip. It was also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex (674699).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Monica Brivio et al.
Analytical chemistry, 77(21), 6852-6856 (2005-11-01)
The reaction of propyl isocyanate (2), benzyl isocyanate (3), and toluene-2,4-diisocyanate (4) with 4-nitro-7-piperazino-2,1,3-benzoxadiazole (1) to yield the corresponding urea derivatives 5 was carried out in a continuous flow glass microfluidics chip. Real-time monitoring of the derivatization reactions was done
Tamara L Church et al.
Journal of the American Chemical Society, 129(26), 8156-8162 (2007-06-15)
The use of mechanistic information to develop a new, catalytic multicomponent reaction is described. The complex [(salph)Al(THF)2]+[Co(CO)4]- (1, salph = N,N'-o-phenylenebis(3,5-di-tert-butylsalicylideneimine), THF = tetrahydrofuran), which is known to carbonylate epoxides, aziridines, and beta-lactones, was used to catalyze the synthesis of
W A Smith et al.
Mutation research, 425(1), 143-152 (1999-03-20)
The large (>2000) and expanding number of natural and synthetic agents with potential cancer chemopreventive properties renders it economically and physically impossible to test each of these agents for their efficacy in the widely accepted 2-year animal bioassay and clinical
Eliana Esparza et al.
Phytochemistry, 179, 112502-112502 (2020-09-02)
Postharvest processing of maca (Lepidium meyenii Walp., Brassicaceae), a traditional high-altitude Andean root crop, involves slow field drying prior to milling into flour. The progressive tissue dehydration and release of hydrolytic enzymes and substrates from cellular compartments results in the
Liang Li et al.
Analytical and bioanalytical chemistry, 411(21), 5465-5479 (2019-06-10)
A benzylureido-β-cyclodextrin was synthesized by the reaction of 6-amino-β-cyclodextrin with an active benzyl isocyanate. Then, it was bonded to silica gel by a thiol-ene addition reaction, obtaining a new benzylureido-β-cyclodextrin-based chiral stationary phase (BzCDP). Its chemical structure was characterized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service