Skip to Content
Merck
All Photos(2)

Key Documents

902810

Sigma-Aldrich

4,4′-Bis(trifluoromethyl)-2,⋅2′-bipyridine

Synonym(s):

Photocatalysis ligand

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$415.00

$415.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$415.00

About This Item

Empirical Formula (Hill Notation):
C12H6F6N2
CAS Number:
Molecular Weight:
292.18
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$415.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

≥95%

form

solid

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: Photocatalysis

mp

78-80 °C

SMILES string

FC(F)(F)c1cc(ncc1)c2nccc(c2)C(F)(F)F

InChI

1S/C12H6F6N2/c13-11(14,15)7-1-3-19-9(5-7)10-6-8(2-4-20-10)12(16,17)18/h1-6H

InChI key

FFOMEQIMPYKURW-UHFFFAOYSA-N

Application

This ligand is used in Ir(III) complexes suitable for use in photocatalysis and phosphorescent OLEDs.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan M O'Donnell et al.
Journal of the American Chemical Society, 138(11), 3891-3903 (2016-02-24)
Excited state proton transfer studies of six Ru polypyridyl compounds with carboxylic acid/carboxylate group(s) revealed that some were photoacids and some were photobases. The compounds [Ru(II)(btfmb)2(LL)](2+), [Ru(II)(dtb)2(LL)](2+), and [Ru(II)(bpy)2(LL)](2+), where bpy is 2,2'-bipyridine, btfmb is 4,4'-(CF3)2-bpy, and dtb is 4,4'-((CH3)3C)2-bpy
Chip Le et al.
Science (New York, N.Y.), 360(6392), 1010-1014 (2018-06-02)
Transition metal-catalyzed arene functionalization has been widely used for molecular synthesis over the past century. In this arena, copper catalysis has long been considered a privileged platform due to the propensity of high-valent copper to undergo reductive elimination with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service