Skip to Content
Merck
All Photos(1)

Key Documents

245550

Sigma-Aldrich

4-Fluorophenylmagnesium bromide solution

2.0 M in diethyl ether

Synonym(s):

p-Fluorophenylmagnesium bromide, Bromo(4-fluorophenyl)magnesium, Bromo(p-fluorophenyl)magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$126.00
800 ML
$447.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$126.00
800 ML
$447.00

About This Item

Linear Formula:
FC6H4MgBr
CAS Number:
Molecular Weight:
199.30
Beilstein:
636802
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

1.043 g/mL at 25 °C

functional group

fluoro

SMILES string

Fc1ccc([Mg]Br)cc1

InChI

1S/C6H4F.BrH.Mg/c7-6-4-2-1-3-5-6;;/h2-5H;1H;/q;;+1/p-1

InChI key

QYBFFRXNNFXREA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Milius et al.
Journal of medicinal chemistry, 34(5), 1728-1731 (1991-05-01)
The synthesis and pharmacological characterization of a series of N-substituted 3-(4-fluorophenyl)tropane derivatives is reported. The compounds displayed binding characteristics that paralleled those of cocaine, and several had substantially higher affinity at cocaine recognition sites. Conjugate addition of 4-fluorophenyl magnesium bromide
S K Aggarwal et al.
Clinical chemistry, 40(8), 1494-1502 (1994-08-01)
A stable isotope dilution gas chromatography-mass spectrometry (GC-MS) method is described for the determination of lead (Pb) in urine and whole blood. The use of lithium bis(trifluoroethyl)dithiocarbamate Pb(FDEDTC) as a chelating agent showed strong memory effect, restricting the range of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service