Skip to Content
Merck
All Photos(1)

Key Documents

331376

Sigma-Aldrich

Phenylmagnesium bromide solution

Phenylmagnesium bromide solution
1 of 1 reviewers received a sample product or took part in a promotion

1.0 M in THF

Synonym(s):

PhMgBr solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$69.00
100 ML
$79.80
4 X 25 ML
$97.50
800 ML
$200.00

$69.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$69.00
100 ML
$79.80
4 X 25 ML
$97.50
800 ML
$200.00

About This Item

Linear Formula:
C6H5MgBr
CAS Number:
Molecular Weight:
181.31
Beilstein:
3588849
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$69.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

density

1.004 g/mL at 25 °C

SMILES string

Br[Mg]c1ccccc1

InChI

1S/C6H5.BrH.Mg/c1-2-4-6-5-3-1;;/h1-5H;1H;/q;;+1/p-1

InChI key

ANRQGKOBLBYXFM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Phenylmagnesium bromide (PhMgBr) solution is an organomagnesium reagent mainly used in Grignard reaction, which finds application as the key step in the synthesis of (S)-2(diphenylmethyl)pyrrolidine, a chiral solvating agent for the NMR analysis.[1] PhMgBr can also undergo palladium-catalyzed Grignard coupling with alkyl chlorides to form the cross-coupled product in good yield.[2]

Other Notes

Storage below 25°C may cause crystallization. Warm gently to redissolve solid.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium?Catalyzed Coupling of Alkyl Chlorides and Grignard Reagents.
Frisch A C, et al.
Angewandte Chemie (International Edition in English), 41(21), 4056-4059 (2002)
A short synthesis of (S)-2-(diphenylmethyl) pyrrolidine, a chiral solvating agent for NMR analysis.
Bailey D J, et al.
Tetrahedron Asymmetry, 8(1), 149-153 (1997)
Ying Li et al.
Analytica chimica acta, 884, 106-113 (2015-06-16)
A novel molecularly imprinted electrochemical sensor based on Fe3O4 nanobeads immobilized on graphene (Fe3O4-MIP@RGO) has been developed for detecting 17β-estradiol (17β-E2) in water using reversible addition fragmentation chain transfer (RAFT) polymerization technique. 17β-E2 can be detected by this electrochemical sensor

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service