Skip to Content
Merck
All Photos(3)

Key Documents

244783

Sigma-Aldrich

Cadmium oxide

powder, 99.5% trace metals basis

Synonym(s):

Cadmia

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$89.90
500 G
$224.00

$89.90

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 13, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$89.90
500 G
$224.00

About This Item

Linear Formula:
CdO
CAS Number:
Molecular Weight:
128.41
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$89.90

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 13, 2025


Request a Bulk Order

Quality Level

Assay

99.5% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: cadmium

density

8.15 g/mL at 25 °C (lit.)

SMILES string

O=[Cd]

InChI

1S/Cd.O

InChI key

CXKCTMHTOKXKQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cadmium oxide (CdO) is a yellowish to reddish-brown inorganic compound known for its semiconductor properties. With a density of 8.15 g/mL at 25 °C, CdO serves as a versatile reagent and catalyst in various chemical reactions. It is primarily used in the production of cadmium-based materials and as a catalyst in organic synthesis. Cadmium oxide is also utilized in the manufacturing of glass and ceramics, where it contributes to color and opacity. Our high-purity cadmium oxide powder, with a purity of 99.5% trace metals basis, is particularly valuable for applications that require minimal impurities.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sung Hun Kim et al.
Nanomaterials (Basel, Switzerland), 10(8) (2020-08-19)
We used low-temperature reactions to synthesize different-sized CdSe quantum dots (QDs) capped with fatty-acid and phosphine ligands. From the correlation of high-resolution transmission electron microscopy and X-ray diffraction (XRD) analyses of the synthesized QDs, we observed size-dependent shape anisotropy. In
Sanjay K Apte et al.
Small (Weinheim an der Bergstrasse, Germany), 7(7), 957-964 (2011-03-10)
The simple, template-free, low-temperature, large-scale synthesis of nanostructured CdS with the hexagonal wurtzite phase from bulk cadmium oxide under solid-phase conditions is demonstrated for the first time. The novel approach involves the homogenization of cadmium oxide (CdO) and thiourea in
Shao-Ying Ting et al.
Optics express, 20(20), 21860-21874 (2012-10-06)
CdZnO/ZnO quantum well (QW) samples are grown on GaN and ZnO templates with plasma-assisted molecular beam epitaxy under different conditions of substrate temperature, Cd effusion cell temperature, and O(2) flow rate for emission characteristics comparison. It is found that the
Oksana V Nesterova et al.
Dalton transactions (Cambridge, England : 2003), (11)(11), 1431-1436 (2008-03-07)
A novel heterometallic 1D coordination polymer [{Ni(en)2}2(micro-NCS)4Cd(NCS)2](n) x nCH3CN (en = ethylenediamine) has been prepared using the self-assembly process in a one-pot reaction of cadmium oxide, nickel and ammonium thiocyanates with an acetonitrile solution of ethylenediamine. The complex consists of
Vahid Safarifard et al.
Ultrasonics sonochemistry, 19(6), 1227-1233 (2012-04-17)
Nanoparticles of a three-dimensional supramolecular Cd(II) compound, [Cd(L)(2)(H(2)O)(2)] (1), (L(-)=1H-1,2,4-triazole-3-carboxylate), have been synthesized by a sonochemical process and characterized by scanning electron microscopy, X-ray powder diffraction, IR spectroscopy and elemental analyses. The thermal stability of compound 1 in both its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service