Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cadmium oxide
Synonym(s):
Cadmium oxide
Linear Formula:
CdO
CAS Number:
1306-19-0
Molecular Weight:
128.41
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Particle Size (µm)
Reagent Type
Core
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
202894
Cadmium oxide,
≥99.99% trace metals basis
Expand
View Pricing
244783
Cadmium oxide,
powder, 99.5% trace metals basis
Expand
View Pricing
1.02015
Cadmium oxide,
EMPLURA
®
Expand
View Pricing