Skip to Content
Merck
All Photos(1)

Documents

1.02015

Supelco

Cadmium oxide

EMPLURA®

Synonym(s):

Cadmium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdO
CAS Number:
Molecular Weight:
128.41
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-146-2
NACRES:
NA.21

Quality Level

product line

EMPLURA®

Assay

≥99.0% (complexometric)

form

solid

potency

72 mg/kg LD50, oral (Rat)

surface area

2.0-4.0 m2/g , BET

particle size

≤5 μm (D 50)

pH

9-10 ( in H2O, aqueous suspension)

mp

950 °C

solubility

0.0021 g/L

density

6.95 g/cm3 at 20 °C

bulk density

400 kg/m3

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Cu: ≤0.001%
Fe: ≤0.001%
Zn: ≤0.005%

storage temp.

2-30°C

InChI

1S/Cd.O

InChI key

CXKCTMHTOKXKQT-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 99.0 %
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.01 %
Fe (Iron): ≤ 0.001 %
Cu (Copper): ≤ 0.001 %
Zn (Zinc): ≤ 0.005 %
Particle size (D 50): ≤ 5.0 µm
Specific surface area (BET): 2.0 - 4.0 m²/g

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service