Skip to Content
Merck
All Photos(1)

Key Documents

224383

Sigma-Aldrich

Isopropylmagnesium chloride solution

2.0 M in diethyl ether

Synonym(s):

Chloro 1-methylethyl magnesium, Chloroisopropylmagnesium, Isopropylmagnesium chloride, iPrMgCl

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$92.10
800 ML
$182.00

$92.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$92.10
800 ML
$182.00

About This Item

Linear Formula:
(CH3)2CHMgCl
CAS Number:
Molecular Weight:
102.85
Beilstein:
4124221
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$92.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

0.834 g/mL at 25 °C

SMILES string

CC(C)[Mg]Cl

InChI

1S/C3H7.ClH.Mg/c1-3-2;;/h3H,1-2H3;1H;/q;;+1/p-1

InChI key

GPIBKUJXKCEZOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The first example of ?-chloromagnesio ?-lactones: their generation from ?-tolylsulfinyl ?-lactones with isopropylmagnesium chloride, stability, and reaction with electrophiles.
Sugiyama S, et al.
Tetrahedron Letters, 47(49), 8771-8775 (2006)
Well-controlled synthesis of block copolythiophenes.
Ohshimizu K and Ueda M.
Macromolecules, 41(14), 5289-5294 (2008)
Synthesis of Poly (3-hexylthiophene) with a Narrower Polydispersity.
Miyakoshi R, et al.
Macromolecular Rapid Communications, 25(19), 1663-1666 (2004)
Preparation of amides mediated by isopropylmagnesium chloride under continuous flow conditions.
Mu?oz JM, et al.
Green Chemistry, 14(5), 1335-1341 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service