Skip to Content
Merck
All Photos(1)

Documents

224391

Sigma-Aldrich

Propylmagnesium chloride solution

2.0 M in diethyl ether

Synonym(s):

n-Propylmagnesium chloride, Chloropropylmagnesium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2MgCl
CAS Number:
Molecular Weight:
102.85
Beilstein:
3600246
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

2.0 M in diethyl ether

density

0.827 g/mL at 25 °C

SMILES string

CCC[Mg]Cl

InChI

1S/C3H7.ClH.Mg/c1-3-2;;/h1,3H2,2H3;1H;/q;;+1/p-1

InChI key

AMXVKMXLDJXAPM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Propylmagnesium chloride solution can be used as a Grignard reagent:
  • For the determination of analytes via derivatization reaction.
  • To synthesize alkyl naphthyl ketones from naphthonitriles.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of n-Alkylnaphthalenes via Semicarbazones
ZENGIN G, et al.
Turkish Journal of Chemistry, 30, 139-144 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service