Skip to Content
Merck
All Photos(1)

Key Documents

218456

Sigma-Aldrich

Ethyl 3-methyl-2-oxobutyrate

97%

Synonym(s):

Ethyl dimethylpyruvate, Ketovaline ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$101.00

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$101.00

About This Item

Linear Formula:
(CH3)2CHCOCOOCH2CH3
CAS Number:
Molecular Weight:
144.17
Beilstein:
1756668
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.410 (lit.)

bp

62 °C/11 mmHg (lit.)

density

0.989 g/mL at 25 °C (lit.)

functional group

ester
ketone

storage temp.

2-8°C

SMILES string

CCOC(=O)C(=O)C(C)C

InChI

1S/C7H12O3/c1-4-10-7(9)6(8)5(2)3/h5H,4H2,1-3H3

InChI key

CKTYYUQUWFEUCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 3-methyl-2-oxobutyrate reacts with a variety of allyl halides in the presence of indium to afford hydroxy unsaturated carbonyl compounds[1]. Conformational behaviour of ethyl 3-methyl-2-oxobutyrate was investigated using solution FTIR in combination with ab initio calculations[2].

Application

Ethyl 3-methyl-2-oxobutyrate was used in the synthesis of (2SR,4SR)-5(S)-(N-Boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid[3].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of (2S, 4S, 5S)-5-(N-BOC)-amino-6-cyclohexyl-4-hydroxy-2-isopropyl-hexanoic acid lactone, an hydroxyethylene dipeptide isostere precusor.
Chakravarty PK, et al.
Tetrahedron Letters, 30(4), 415-418 (1989)
Conformational isomerism of a-ketoesters. A FTIR and ab initio study.
Ferri D, et al.
J. Chem. Soc. Perkin Trans. II, 2, 221-227 (2000)
A convenient allylation of 1, n-dicarbonyl compounds using organoindium reagents.
Lee PH, et al.
Bull. Korean Chem. Soc., 22(12), 1380-1384 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service