Skip to Content
Merck
All Photos(1)

Documents

75950

Sigma-Aldrich

2-Oxovaleric acid

≥98.0% (T)

Synonym(s):

α-Ketovaleric acid, 2-Oxopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COCOOH
CAS Number:
Molecular Weight:
116.12
Beilstein:
635884
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (T)

form

liquid

bp

88-90 °C/12 mmHg (lit.)

mp

7-9 °C (lit.)

density

1.110 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCC(=O)C(O)=O

InChI

1S/C5H8O3/c1-2-3-4(6)5(7)8/h2-3H2,1H3,(H,7,8)

InChI key

KDVFRMMRZOCFLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tara Kearney et al.
Metabolism: clinical and experimental, 52(1), 50-59 (2003-01-14)
In this study, we concurrently examined the effects of 8 and 40 weeks of growth hormone replacement (GHR) on lipids, lipoprotein composition, low-density lipoprotein (LDL) size, very-low-density lipoprotein (VLDL) apolipoprotein (apo)B kinetics and LDL apoB kinetics. Eight weeks of GHR
R Sakowicz et al.
Biochemistry, 32(47), 12730-12735 (1993-11-30)
Threonine 246 is an active site residue that is conserved in all known L-lactate dehydrogenase (LDH; EC 1.1.1.27) sequences. In order to investigate the role of Thr246 in Bacillus stearothermophilus LDH, this residue was altered by site-directed mutagenesis to valine
C Coronel et al.
Molecular and biochemical parasitology, 4(1-2), 29-38 (1981-11-01)
Whole cell extracts of culture epimastigotes of Trypanosoma cruzi (Tulahuén strain) have alpha-hydroxyacid dehydrogenase activity which catalyzes the NAD-linked reaction alpha-ketoacid in equilibrium with alpha-hydroxyacid, with a variety of substrates. Two molecular forms of the enzyme have been separated by
Carmen Martínez-Cuesta et al.
FEMS microbiology letters, 217(1), 109-113 (2002-11-26)
The effect of the bacteriocin lacticin 3147 on the branched-chain amino acid transamination by Lactococcus lactis IFPL359 was investigated. The bacteriocin provokes membrane permeabilisation of the cells, rendering them non-viable but metabolically active. Free diffusion of amino acids into the
R Bränström et al.
The Journal of biological chemistry, 273(23), 14113-14118 (1998-06-11)
The ATP-regulated potassium (KATP) channel plays an essential role in the control of insulin release from the pancreatic beta-cell. In the present study we have used the patch-clamp technique to study the direct effects of alpha-ketoisocaproate on the KATP channel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service