Skip to Content
Merck
All Photos(2)

Documents

E47808

Sigma-Aldrich

Ethyl pyruvate

98%

Synonym(s):

2-Oxopropanoic acid ethyl ester, Ethyl 2-oxopropanoate, Ethyl 2-oxopropionate, Ethyl methylglyoxylate, Methyl ethoxycarbonyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOOC2H5
CAS Number:
Molecular Weight:
116.12
Beilstein:
1071466
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.404 (lit.)

bp

144 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C(C)=O

InChI

1S/C5H8O3/c1-3-8-5(7)4(2)6/h3H2,1-2H3

InChI key

XXRCUYVCPSWGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Ethyl pyruvate can undergo asymmetric Henry reaction with nitromethane to form α-hydroxy β-nitro esters. It can be used as the model compound for α-ketoesters to study the mechanism of enantioselective hydrogenation reactions.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Enantioselective Henry (Nitroaldol) Reaction of Aldehydes and ?-Ketoesters Catalyzed by N,N'-Dioxide-Copper (I) Complexes.
Qin B, et al.
The Journal of Organic Chemistry, 72(24), 9323-9328 (2007)
Lin Sun et al.
Journal of neurotrauma, 36(3), 421-435 (2018-06-23)
High mobility group box-1 (HMGB1) could function as an early trigger for pro-inflammatory activation after spinal cord injury (SCI). Spinal cord edema contributes to inflammatory response mechanisms and a poor clinical prognosis after SCI, for which efficient therapies targeting the
Runkuan Yang et al.
Critical care (London, England), 16(1), R9-R9 (2012-01-18)
Inflammation may critically affect mechanisms of liver injury in acetaminophen (APAP) hepatotoxicity. Kupffer cells (KC) play important roles in inflammation, and KC depletion confers protection at early time points after APAP treatment but can lead to more severe injury at
98% Enantioselectivity in the asymmetric synthesis of a useful chiral building block by heterogeneous method: Enantioselective hydrogenation of ethyl-benzoylformate over cinchona modified Pt/Al2 O3 catalysts in the acetic acid.
Sutyinszki M, et al.
Catalysis Communications, 3(3), 125-127 (2002)
M P Fink
Journal of internal medicine, 261(4), 349-362 (2007-03-30)
Ethyl pyruvate (EP) is a simple derivative of the endogenous metabolite, pyruvic acid. Treatment with EP has been shown to improve survival and/or ameliorate organ dysfunction in a wide variety of preclinical models of critical illnesses, such as severe sepsis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service