Skip to Content
Merck
All Photos(3)

Documents

215945

Sigma-Aldrich

Pararosaniline acetate

Dye content 90 %

Synonym(s):

Basic Parafuchsin, Basic Red 9, Magenta O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3 · C2H4O2
CAS Number:
Molecular Weight:
347.41
Colour Index Number:
42500
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

solid

composition

Dye content, 90%

mp

203 °C (dec.) (lit.)

solubility

95% ethanol: 0.1%

λmax

545 nm

ε (extinction coefficient)

≥11000 at 235-241 nm in ethanol: water (1:1) at 0.004%
≥16000 at 286-292 nm in ethanol: water (1:1) at 0.004%
≥78000 at 543-549 nm in ethanol: water (1:1) at 0.004%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(O)=O.Nc1ccc(cc1)C(\c2ccc(N)cc2)=C3\C=CC(=N)C=C3

InChI

1S/C19H17N3.C2H4O2/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;1-2(3)4/h1-12,20H,21-22H2;1H3,(H,3,4)

InChI key

YIXIVOYGLPFDCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pararosaniline acetate is one of the components of basic fuchsin. It acts as a reversible, linear inhibitor of horse butyrylcholinesterase (BChE).

Other Notes

This is the pure, principal component of mixtures commonly referred to as Basic Fuchsin.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D. Larra?aga, Richard J. Lewis, Robert A. Lewis
Hawley's Condensed Chemical Dictionary (2016)
Comparative effects of cationic triarylmethane, phenoxazine and phenothiazine dyes on horse serum butyrylcholinesterase.
Yucel YY
Archives of Biochemistry and Biophysics, 478, 201-205 (2008)
C.I. Basic Red 9 monohydrochloride.
Report on carcinogens : carcinogen profiles, 11, III66-III67 (2004-01-01)
M E Baker et al.
The Analyst, 119(5), 959-961 (1994-05-01)
The colorimetric indicator pararosaniline has been immobilized onto the cation-exchange resins Amberlite IRC-50, Dowex 50W-X8 and cellulose phosphate by electrostatic bonding. The reflectance of each reagent phase was measured using a bifurcated fibre-optic system and a flow cell. Pararosaniline immobilized
Determination of formaldehyde in indoor air by a solid sorbent technique.
P E Georghiou et al.
IARC scientific publications, (109)(109), 251-255 (1993-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service