If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
1.88052
Water Standard
1%, Certified Reference Material for volumetric Karl Fischer Titration 1 g ≙ 10 mg H₂O Aquastar®
Synonym(s):
Water standard 1% for volumetric Karl Fischer Titration
About This Item
Recommended Products
Product Name
Water standard 1%, Certified Reference Material for volumetric Karl Fischer Titration 1 g ≙ 10 mg H₂O Aquastar®
grade
certified reference material
Quality Level
Agency
ISO (17025, 17034)
product line
Aquastar®
form
liquid
autoignition temp.
287 °C (DIN 51794,Information refers to the main component.)
expl. lim.
1.7 % (v/v)
technique(s)
Karl-Fischer titration: suitable
titration: suitable
bp
119 °C/1013 hPa
mp
-95 °C
transition temp
flash point 31 °C (Information refers to the main component.)
density
0.92 g/cm3 at 20 °C
format
single component solution
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
- Evaluating the accuracy of coulometric Karl Fischer titrators according to ISO 9001 chapter 7.1.5 “Monitoring and measuring resources” as a Certified reference material (CRM)
- Checking the accuracy of measured results according to European Pharmacopeia (Ph.Eur.) chapter 2.5.32 “Water Micro Determination”
- Standardizing titrants according to European Pharmacopeia (Ph.Eur.) chapter 2.5.12 “Water Semi-Micro Determination” and the United States Pharmacopeia <921> “Water Determination”
- Titer determinations using volumetric method with one component and two component reagents
Features and Benefits
- Packed in convenient 8 mL glass ampoules and 10 single-use ampoules in a box
- Supplied with a batch-specific certificate of analysis (CoA) having data on the exact measured water content of the batch, uncertainty, method of analysis, the NIST batch, and minimum shelf life
- Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited calibration laboratory
- Directly traceable to SI unit (kg) and verified by SRM 2890 from NIST
Packaging
Analysis Note
Legal Information
reagent
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
87.8 °F - Information refers to the main ingredient.
Flash Point(C)
31 °C - Information refers to the main ingredient.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Methods including internal extraction, external extraction, and Karl Fischer oven ensure accurate water content determination by titration.
SmartChemicals enable wireless CoA data transfer, reducing human error in titration experiments.
Protocols
This protocol describes a one-component Volumetric Karl Fischer Titration method for the determination of water content in Benserazide hydrochloride.
Foodstuffs often provide a challenge for the water determination by Karl-Fischer. Instant Coffee is such a sample matrix and its water content has influence on quality and shelf life.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
Can you please confirm difference between these two products with SKU number 1880520010 and 1880520013 because their description is same.
1 answer-
Both products are the same. The only difference is that product 1880520313 is a "SmartChemical" with an RFID tag on the label for seamless data transfer to the instrument. Visit www.sigmaaldrich.com/SmartChemicals<http://www.sigmaaldrich.com/SmartChemicals> for additional information.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service