Skip to Content
Merck
All Photos(1)

Key Documents

215082

Sigma-Aldrich

trans-1,4-Dichloro-2-butene

technical grade, 85%, remainder predominantly cis isomer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH=CHCH2Cl
CAS Number:
Molecular Weight:
125.00
Beilstein:
1719693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical grade

Quality Level

vapor pressure

10 mmHg ( 20 °C)

Assay

85%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

74-76 °C/40 mmHg (lit.)

mp

1-3 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC\C=C\CCl

InChI

1S/C4H6Cl2/c5-3-1-2-4-6/h1-2H,3-4H2/b2-1+

InChI key

FQDIANVAWVHZIR-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-1,4-Dichloro-2-butene was used as starting reagent in the synthesis of trans-tetrahydrofuran (THF) ring building block[1]. It was also employed in the chemical synthesis of swainsonine [(1S,2R,8R,8 αR)-trihydroxyindolizidine][2] and 9-alkylpurines[3].

Other Notes

85%, remainder predominantly cis isomer

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Elbein et al.
Biochemistry, 26(9), 2502-2510 (1987-05-05)
The chemical synthesis of swainsonine [(1S,2R,8R,8 alpha R)-trihydroxyindolizidine] from trans-1,4-dichloro-2-butene was previously described [Adams, C. E., Walker, F. J., & Sharpless, K. B. (1985) J. Org. Chem. 50, 420-424]. A modification of that synthesis provided two other isomers, referred to
Z M Wang et al.
Chirality, 12(7), 581-589 (2000-06-22)
A highly efficient synthetic method for the trans-tetrahydrofuran (THF) ring building block was established and the title compound was synthesized in 19 steps from trans-1,4-dichloro-2-butene via a convergent route with a Wittig reaction as the key step.
S Phadtare et al.
Journal of medicinal chemistry, 30(2), 437-440 (1987-02-01)
Alkylation of adenine (5a) or 2-amino-6-chloropurine (5b) with excess trans-1,4-dichloro-2-butene (4), effected by K2CO3 in dimethyl sulfoxide or tetra-n-butylammonium fluoride in tetrahydrofuran, led in 90-95% regioselectivity to 9-alkylpurines 6a and 6b. The title compounds 2a and 2b were obtained by
R J Gardner et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 23(1), 87-92 (1985-01-01)
Several important chemicals, including formaldehyde, 1,4-dichloro-2-butene, bis-chloromethyl ether, hexamethylphosphoramide, and epichlorohydrin have been shown to produce nasal tumours in rats following repeated or continuous inhalation exposures. Some of these compounds are respiratory irritants. To determine whether there is a correlation
L S Mullin et al.
Drug and chemical toxicology, 25(2), 227-230 (2002-05-25)
Rats were exposed by inhalation to either 0.5 ppm 1,4-dichlorobutene-2 (DCB) for two years or to 5.0 ppm for seven months, 2.5 ppm for five months, and no further exposure for 12 months prior to sacrifice. Malignant and non-malignant tumors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service