196193
Red-Al® sodium bis(2-methoxyethoxy)aluminum hydride solution
≥60 wt. % in toluene
Synonym(s):
SBAH, Sodium bis(2-methoxyethoxy)aluminum dihydride, Sodium dihydrido-bis(2-methoxyethoxy)aluminate
About This Item
Recommended Products
vapor pressure
21 mmHg ( 20 °C)
Quality Level
expl. lim.
7 %
reaction suitability
reagent type: reductant
concentration
≥60 wt. % in toluene
density
1.036 g/mL at 25 °C
SMILES string
[Na+].[H][Al-]([H])(OCCOC)OCCOC
InChI
1S/2C3H7O2.Al.Na.2H/c2*1-5-3-2-4;;;;/h2*2-3H2,1H3;;;;/q2*-1;2*+1;;
InChI key
XJIQVZMZXHEYOY-UHFFFAOYSA-N
General description
- Aldehydes, ketones, esters, and anhydrides to primary alcohols.
- Ketoximes and aldoximes to primary amines.
- Cyclic compounds such as lactones and epoxides to diols.
It is also used as a catalyst in the ring-opening polymerization reactions.
SBAH is a reducing agent used to reduce functional groups like carbonyls, amides, epoxides, esters, and carboxylic acids.
Application
- A tosyl deprotecting agent.
- A catalyst for crosslinking of polyvinylsilanes (PVS) by Si-Si dehydrocoupling reaction in the presence of group 4 metallocene complexes.
- A reducing reagent for the synthesis of optically active N-protected amino alcohols and peptide alcohols.
Legal Information
Signal Word
Danger
Hazard Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
39.2 °F - closed cup
Flash Point(C)
4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Red-Al® is a versatile and safe reducing agent, serving as a substitute for LiAlH4 in many reactions, with nonpyrophoric properties.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service