Skip to Content
Merck
All Photos(1)

Key Documents

209716

Sigma-Aldrich

Ethyl (methylthio)acetate

98%

Synonym(s):

(Methylthio)acetic acid ethyl ester, Ethyl α-(methylthio)acetate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$108.00

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$108.00

About This Item

Linear Formula:
CH3SCH2CO2C2H5
CAS Number:
Molecular Weight:
134.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.459 (lit.)

bp

70-72 °C/25 mmHg (lit.)

density

1.043 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CSC

InChI

1S/C5H10O2S/c1-3-7-5(6)4-8-2/h3-4H2,1-2H3

InChI key

MDIAKIHKBBNYHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl (methylthio)acetate is an important sulphur-containing aroma compound isolated from melon Cucumis melo L[1].

Application

Ethyl (methylthio)acetate was used as internal standard during the analytical determination of volatile sulfur compounds in port wines[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfur volatiles in Cucumis melo cv. Makdimon (Muskmelon) aroma.
Wyllie SG, et al.
Sulfur Compounds in Foods, 36-48 (1994)
António César Silva Ferreira et al.
Journal of agricultural and food chemistry, 51(3), 727-732 (2003-01-23)
Volatile sulfur compounds of 15 young port wines and 12 old port wines were determined. As there is a great difference in the pool of sulfur compounds between the two groups of wines, an experimental protocol was performed to determine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service