Skip to Content
Merck
All Photos(1)

Key Documents

W400301

Sigma-Aldrich

Methyl (methylthio)acetate

FG

Synonym(s):

Methyl (methylmercapto)acetate, Methyl S-methylthioglycolate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$237.00
100 G
$861.00

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
25 G
$237.00
100 G
$861.00

About This Item

Linear Formula:
CH3SCH2COOCH3
CAS Number:
Molecular Weight:
120.17
FEMA Number:
4003
Beilstein:
1902431
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.146
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

impurities

may contain ≤50 ppm Methyl chloroacetate (residual solvent)

refractive index

n20/D 1.4650 (lit.)

bp

49-51 °C/12 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

nutty; roasted

storage temp.

2-8°C

SMILES string

COC(=O)CSC

InChI

1S/C4H8O2S/c1-6-4(5)3-7-2/h3H2,1-2H3

InChI key

HZYCAKGEXXKCDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service