Skip to Content
Merck
All Photos(1)

Documents

W328200

Sigma-Aldrich

Ethyl thioacetate

≥98%

Synonym(s):

S-Ethyl acetothioate, S-Ethyl ethanethioate, S-Ethyl thioacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COSC2H5
CAS Number:
Molecular Weight:
104.17
FEMA Number:
3282
EC Number:
Council of Europe no.:
11665
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.018
NACRES:
NA.21

biological source

synthetic

Agency

meets purity specifications of JECFA

Assay

≥98%

refractive index

n20/D 1.458 (lit.)

bp

116 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

garlic; onion; fruity; sulfurous

SMILES string

CCSC(C)=O

InChI

1S/C4H8OS/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

APTGPWJUOYMUCE-UHFFFAOYSA-N

Related Categories

General description

Ethyl thioacetate is a volatile sulfur compound that is reported to occur in durian fruit and wine.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of low-volatility organic sulphur compounds in wines by solid-phase microextraction and gas chromatography.
Mestres M, et al.
Journal of Chromatography A, 881(1), 583-590 (2000)
Marlize Z Bekker et al.
Food chemistry, 207, 148-156 (2016-04-16)
The effects of pH and Cu(2+) treatment on the formation of volatile sulfur compounds (VSCs) were investigated in Chardonnay and Shiraz wine samples. Four VSCs were significantly affected by pH, with lower wine pH associated with decreased hydrogen sulfide (H2S)
Antonio G Cordente et al.
Applied and environmental microbiology, 85(6) (2019-01-20)
During alcoholic fermentation of grape sugars, wine yeasts produce a range of secondary metabolites that play an important role in the aroma profile of wines. In this study, we have explored the ability of a large number of wine yeast
Sulfur-containing volatiles of durian fruits (Durio zibethinus Murr.).
Weenen H, et al.
Journal of Agricultural and Food Chemistry, 44(10), 3291-3293 (1996)
Stéphanie Rollero et al.
Food microbiology, 94, 103650-103650 (2020-12-07)
The positive impact of certain non-Saccharomyces yeasts on the aromatic profile of wines has been well documented in literature and their industrial use in association with S. cerevisiae is now recommended. Competition between non-Saccharomyces species and Saccharomyces cerevisiae for various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service