Skip to Content
Merck
All Photos(2)

Key Documents

205028

Sigma-Aldrich

Zirconyl chloride hydrate

99.99% trace metals basis

Synonym(s):

Zirconium Dichloride Oxide Hydrate, Zirconium oxychloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$127.00
50 G
$379.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$127.00
50 G
$379.00

About This Item

Linear Formula:
ZrOCl2·xH2O
CAS Number:
Molecular Weight:
178.13 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

99.99% trace metals basis

form

solid

composition

Degree of hydration, 6-8

impurities

≤150.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

SMILES string

O.Cl[Zr](Cl)=O

InChI

1S/2ClH.H2O.O.Zr/h2*1H;1H2;;/q;;;;+2/p-2

InChI key

SDFOCNSOSARAKC-UHFFFAOYSA-L

Application

Building block for a new series of mixed zirconium phosphonates which may have uses in catalysis, ion exchange or photophysics.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefania Vitale et al.
Nanoscale, 11(11), 4788-4793 (2019-01-31)
The fabrication of stable, highly conductive molecular nano-junctions is one of the main research goals in the field of molecular electronics. In this paper we report on the self-assembly and functional characterisation of highly conductive molecular wires, based on mixed-metal
Leenstra, W.R. Amicangelo, J.C.
Inorganic Chemistry, 37, 5317-5317 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service