Skip to Content
Merck
All Photos(3)

Key Documents

380679

Sigma-Aldrich

Zirconium(IV) oxynitrate hydrate

99.99% trace metals basis

Synonym(s):

Zirconyl nitrate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$156.00
50 G
$555.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$156.00
50 G
$555.00

About This Item

Linear Formula:
ZrO(NO3)2 · xH2O
CAS Number:
Molecular Weight:
231.23 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

99.99% trace metals basis

form

powder

composition

Degree of hydration, ~1

impurities

≤150.0 ppm Trace Metal Analysis

application(s)

battery manufacturing

SMILES string

O=[Zr++].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.H2O.O.Zr/c2*2-1(3)4;;;/h;;1H2;;/q2*-1;;;+2

InChI key

BGMQNYCWALSARE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Indoles and aldehydes can undergo condensation reaction in the presence of zirconium (IV) oxynitrate hydrate to form bis- or tris(indolyl)methane products, which can undergo dehydrogenation to yield conjugated chromophores to be used as pH indicators (pH range 8–10).[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zirconium(IV)-catalyzed one-pot synthesis and oxidation of bis- and tris(indolyl)methanes into conjugated chromophores as new pH indicators or calorimetric chemosensors for transition metals
Khorshidi A, et al.
Tetrahedron Letters, 55(29), 3873-3877 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service