Skip to Content
Merck
All Photos(3)

Key Documents

200220

Sigma-Aldrich

Azure II eosinate

powder

Synonym(s):

Azure II-eosin; Methylene blue

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$500.00
100 G
$1,620.00

$500.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$500.00
100 G
$1,620.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$500.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

technique(s)

microbe id | staining: suitable

color

dark green

solubility

soluble, hazy to turbid

λmax

524 nm (2nd)
647 nm

Amax

≥0.50 at 645-655 nm in methanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Cl-].[Cl-].CNc1ccc2N=C3C=C\C(C=C3Sc2c1)=[N+](/C)C.CN(C)c4ccc5N=C6C=C\C(C=C6Sc5c4)=[N+](/C)C.[O-]C(=O)c7ccccc7C8=C9C=C(Br)C(=O)C(Br)=C9Oc%10c(Br)c([O-])c(Br)cc8%10

InChI

1S/C20H8Br4O5.C16H18N3S.C15H15N3S.2ClH.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;;;;/h1-6,25H,(H,27,28);5-10H,1-4H3;4-9H,1-3H3;2*1H;;/q;+1;;;;2*+1/p-3

InChI key

CSDDIABEPMTRIP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Chief ingredient of Giemsa Stain.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service