Skip to Content
Merck
All Photos(2)

Key Documents

200239

Sigma-Aldrich

Azure B eosinate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$403.00

$403.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$403.00

About This Item

Linear Formula:
C20H6Br4O5 ⋅ 2C15H16N3S
CAS Number:
Molecular Weight:
1186.62
EC Number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$403.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder or crystals

solubility

acetic acid: 1 mg/mL

λmax

523 nm (2nd)
636 nm

ε (extinction coefficient)

≥142400 at 635-641 nm in methanol at 0.005 g/L
≥47500 at 241-247 nm in methanol at 0.005 g/L
≥82000 at 521-527 nm in methanol at 0.005 g/L
≥94900 at 286-292 nm in methanol at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

BrC1=CC2=C(C(Br)=C1[O-])OC3=C(C=C(Br)C([O-])=C3Br)C24OC(C5=C4C=CC=C5)=O.C[N+](C)=C(C=C6)C=C(C6=N7)SC8=C7C=CC(NC)=C8.C[N+](C)=C(C=C9)C=C(C9=N%10)SC%11=C%10C=CC(NC)=C%11

InChI

1S/C20H8Br4O5.2C15H15N3S/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20;2*1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12/h1-6,25-26H;2*4-9H,1-3H3

InChI key

NCBQBFXQRBYYGY-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jordan R Jensen et al.
Bioorganic & medicinal chemistry, 19(17), 5147-5154 (2011-08-02)
Whole brain imaging of tau-bearing neurofibrillary lesions has the potential to improve the premortem diagnosis and staging of Alzheimer's disease. Diverse compounds with high affinity for tau aggregates have been reported from high-throughput screens, but the affinity driving features common

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service