Skip to Content
Merck
All Photos(1)

Key Documents

196517

Sigma-Aldrich

9-(Chloromethyl)anthracene

≥98%

Synonym(s):

9-Anthracenylmethyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl
CAS Number:
Molecular Weight:
226.70
Beilstein:
1873394
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥98%

mp

138-140 °C (lit.)

SMILES string

ClCc1c2ccccc2cc3ccccc13

InChI

1S/C15H11Cl/c16-10-15-13-7-3-1-5-11(13)9-12-6-2-4-8-14(12)15/h1-9H,10H2

InChI key

PCVRSXXPGXRVEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Protecting reagent for carboxylic acids, phenols, thiophenols, and mercaptans.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A significant fluorescence quenching of anthrylaminobenzocrown ethers by paramagnetic metal cations
Chang, J. H., Choi, Y. M., & Shin, Y. K.
Bull. Korean Chem. Soc., 22(5), 527-530 (2001)
Dorothea F K Rawn et al.
Journal of chromatography. A, 1080(2), 148-156 (2005-07-13)
A rapid and simple method for confirmation of the diarrhetic shellfish poisons (DSP): okadaic acid (OA), dinophysistoxin-1 (DTX-1) and dinophysistoxin-2 (DTX-2) using fluorescence detection following derivatization with 9-chloromethylanthracene, has been established as an alternate to LC/MS. Exposure of the anthrylmethyl
Clickable Poly (2-Oxazoline) s as Versatile Building Blocks.
Fijten, M. W., Haensch, C., van Lankvelt, B. M., Hoogenboom, R., & Schubert, U. S.
Macromolecular Chemistry and Physics, 209(18), 1887-1895 (2008)
9 (chloromethyl) anthracene: a useful derivatizing reagent for enhanced ultraviolet and fluorescence detection of carboxylic acids with liquid chromatography
Korte, W. D
Journal of Chromatography A, 243(1), 153-157 (1982)
J F Lawrence et al.
Journal of chromatography. A, 721(2), 359-364 (1996-01-19)
The reagent 9-chloromethylanthracene was evaluated for derivatization of the diarrhetic shellfish poisons, okadaic acid and dinophysistoxin-1 (DTX-1), to form fluorescent products separable by liquid chromatography. The toxins were reacted with the reagent in acetonitrile in the presence of tetramethylammonium hydroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service