Skip to Content
Merck
All Photos(1)

Key Documents

857270

Sigma-Aldrich

(+)-Cinchonine

85%

Synonym(s):

Cinchonine monohydrochloride dihydrate, NSC 6176

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$89.10
100 G
$204.00

$89.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$89.10
100 G
$204.00

About This Item

Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39
Beilstein:
89689
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

$89.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

85%

form

solid

optical activity

[α]23/D +228°, c = 0.5 in ethanol

mp

258-260 °C (lit.)

functional group

hydroxyl

SMILES string

[H][C@@]12CCN(C[C@H]1C=C)[C@]([H])(C2)[C@@H](O)c3ccnc4ccccc34

InChI

1S/C19H22N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h2-7,9,13-14,18-19,22H,1,8,10-12H2/t13-,14-,18+,19-/m0/s1

InChI key

KMPWYEUPVWOPIM-QAMTZSDWSA-N

General description

(+)-Cinchonine, one of the alkaloids found in the barks of cinchona tree,[1] is mainly used in the treatment of malaria.[2] It belongs to the monoclinic crystal system and P21 space group.[3] The solubility of cinchonine can be improved by the formation of inclusion complexes with cyclodextrins.[2]

Application

(+)-Cinchonine, in the presence of lithium diisopropylamide (LDA) forms a complex, which can catalyze the asymmetric conjugate addition of benzyl- and alkylphosphonates to aromatic and heteroaromatic nitroalkenes to form the corresponding adducts.[4]

Other Notes

remainder dihydrocinchonine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Cinchonine catalyzed diastereo-and enantioselective Michael addition of a-lithiated phosphonates to nitroalkenes.
Rai V, et al.
Tetrahedron Asymmetry, 18(22), 2719-2726 (2007)
Song CE.
Cinchona Alkaloids in Synthesis and Catalysis: Ligands, Immobilization and Organocatalysis, 2-3 (2009)
The molecular and crystal structure of the alkaloid cinchonine.
Oleksyn B, et al.
Acta Crystallographica Section B, Structural Science, 35(2), 440-444 (1979)
Chensong Pan et al.
Journal of the American Society for Mass Spectrometry, 16(2), 263-270 (2005-02-08)
A method with carbon nanotubes functioning both as the adsorbent of solid-phase extraction (SPE) and the matrix for matrix assisted laser desorption/ionization mass spectrometry (MALDI-MS) to analyze small molecules in solution has been developed. In this method, 10 microL suspensions
Yu-Hua Liao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(21), 6679-6687 (2012-04-14)
An asymmetric conjugate addition of 3-monosubstituted oxindoles to a range of (E)-1,4-diaryl-2-buten-1,4-diones, catalyzed by commercially available cinchonine, is described. This organocatalytic asymmetric reaction affords a broad range of 3,3'-disubstituted oxindoles that contain a 1,4-dicarbonyl moiety and vicinal quaternary and tertiary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service