Skip to Content
Merck
All Photos(1)

Key Documents

758388

Sigma-Aldrich

Pyrazino[2,3-f][1,10]phenanthroline

99% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$297.00

$297.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$297.00

About This Item

Empirical Formula (Hill Notation):
C14H8N4
CAS Number:
Molecular Weight:
232.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$297.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99% (HPLC)

form

solid

mp

310-315 °C

SMILES string

c1cnc2c(c1)c3nccnc3c4cccnc24

InChI

1S/C14H8N4/c1-3-9-11(15-5-1)12-10(4-2-6-16-12)14-13(9)17-7-8-18-14/h1-8H

InChI key

IBOSPAWVGHGUAV-UHFFFAOYSA-N

General description

Pyrazino[2,3-f][1,10]phenanthroline is a ligand which is used in the synthesis of copper(I)- phenanthroline complexes which are used in molecular device technology and in solar-energy conversion. It functions as an electron acceptor and the electron density is delocalized on the quinoxaline ring. The absorption and emission properties and the excited state lifetimes of these phenanthroline based copper complexes can be fine-tuned easily.[1]

Application

  • As an electron accepting ligand in metal complexes which are used in solar cells.[1]
  • Used in the synthesis of ruthenium (II) complexes which are used as DNA probes.[2]

Features and Benefits

Imparts improved directionality of electron transfer in excited copper complexes.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujie Sun et al.
Inorganic chemistry, 47(14), 6427-6434 (2008-06-25)
A series of ruthenium(II) complexes possessing ligands with an extended pi system were synthesized and characterized. The complexes are derived from [Ru(bpy)3](2+) (1, bpy = 2,2'-bipyridine) and include [Ru(bpy)2(tpphz)](2+) (2, tpphz = tetrapyrido[3,2-a:2',3'-c:3'',2''-h:2''',3'''-j]phenazine), [Ru(bpy)2(dppx)](2+) (3, dppx = 7,8-dimethyldipyrido[3,2-a:2',3'-c]phenazine), [Ru(bpy)2(dppm2)](2+) (4
Synthesis, spectroscopic and electrochemical characterization of copper (I) complexes with functionalized pyrazino [2, 3-f]-1, 10-phenanthroline.
Barrientos, L., Araneda, C., Loeb, B., & Crivelli, I. G.
Polyhedron, 27(4), 1287-1295 (2008)
Ge; Hu; Wei;et al.
Journal of Luminescence, 131, 238-243 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service