All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H13FN2
CAS Number:
Molecular Weight:
180.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
solid
bp
118-123 °C/0.1 mmHg (lit.)
mp
30-33 °C (lit.)
SMILES string
Fc1ccc(cc1)N2CCNCC2
InChI
1S/C10H13FN2/c11-9-1-3-10(4-2-9)13-7-5-12-6-8-13/h1-4,12H,5-8H2
InChI key
AVJKDKWRVSSJPK-UHFFFAOYSA-N
Gene Information
rat ... Chrnb2(54239)
Application
1-(4-Fluorophenyl)piperazine was used in the synthesis of N,N-disubstituted piperazine.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Craig A Stockmeier et al.
Journal of psychiatric research, 43(10), 887-894 (2009-02-14)
Serotonin-1A receptors may play a role in the pathophysiology of depression and suicide. In postmortem brain tissue, agonist binding to serotonin-1A receptors is reportedly increased or unchanged in depression or suicide, while neuroimaging studies report a decrease in antagonist binding
P E Keane et al.
Neuropharmacology, 21(2), 163-169 (1982-02-01)
Niaprazine (60 mg/kg i.p.) increased rat brain 5-hydroxyindole acetic acid (5-HIAA) concentrations 30 min after treatment, and reduced them at 3-8 hr after treatment. Rat brain 5-hydroxytryptamine (5-HT) levels were unchanged. Niaprazine also produced a short-lasting depletion of rat brain
Rapid synthesis of N, N'-disubstituted piperazines. Application to the preparation of No carrier added 1-(4-[18F] fluorophenyl) piperazine and of an [18F]-selective ligand of serotoninergic receptors (5HT2 antagonist).
Collins M, et al.
Journal of the Chemical Society. Perkin Transactions 1, 23, 3185-3188 (1992)
Camilla Montesano et al.
Drug testing and analysis, 9(5), 798-807 (2016-07-28)
In this paper, an analytical method has been developed and validated for the analysis of new psychoactive substances (NPS) and metabolites in hair samples. The method was based on pressurized liquid extraction (PLE) followed by solid-phase extraction (SPE) clean-up and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service