Skip to Content
Merck
All Photos(1)

Key Documents

73659

Sigma-Aldrich

1-(4-Chlorophenyl)piperazine

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$63.00
5 G
$145.00

$63.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$63.00
5 G
$145.00

About This Item

Empirical Formula (Hill Notation):
C10H13ClN2
CAS Number:
Molecular Weight:
196.68
Beilstein:
150981
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$63.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Assay

≥98.0% (GC)

form

crystals

mp

76-79 °C

functional group

chloro

SMILES string

Clc1ccc(cc1)N2CCNCC2

InChI

1S/C10H13ClN2/c11-9-1-3-10(4-2-9)13-7-5-12-6-8-13/h1-4,12H,5-8H2

InChI key

UNEIHNMKASENIG-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Maj et al.
Polish journal of pharmacology and pharmacy, 32(4), 495-504 (1980-07-01)
Studies of 1-(m-chlorophenyl)-piperazine (m-CPP), 1-(p-chlorophenyl)-piperazine (p-CPP) and 1-phenylpiperazine (PP) were carried out on rats, mice and rabbits in order to assess their stimulatory effect on the central serotonin system. It was found out that m-CPP and p-CPP evoked a characteristic
R W Fuller et al.
Research communications in chemical pathology and pharmacology, 29(1), 201-204 (1980-07-01)
1-(p-Chlorophenyl)piperazine (PCPP), a structural analog of p-chloroamphetamine (PCA), increased serotonin and decreased 5-hydroxyindoleacetic acid (5-HIAA) concentration in rat brain at 6 hours. At one day and one week, serotonin and 5-HIAA concentrations were normal. In contrast, PCA lowered serotonin and
Piotr Cysewski
Journal of molecular modeling, 23(4), 136-136 (2017-03-30)
In silico screening was performed to search for binary solids in which a phenylpiperazine-derivative drug was cocrystallized with a dicarboxylic acid. The phenylpiperazine derivative could be any of 61 such drugs, while the dicarboxylic acid could be any of nine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service