Skip to Content
Merck
All Photos(2)

Key Documents

170275

Sigma-Aldrich

2-Methyl-1-phenyl-2-propanol

98%

Synonym(s):

1,1-Dimethyl-2-phenylethyl alcohol, Benzyl dimethyl carbinol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$74.40

$74.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$74.40

About This Item

Linear Formula:
C6H5CH2C(CH3)2OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1855608
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

refractive index

n20/D 1.514 (lit.)

bp

94-96 °C/10 mmHg (lit.)

mp

23-25 °C (lit.)

density

0.974 g/mL at 25 °C (lit.)

SMILES string

CC(C)(O)Cc1ccccc1

InChI

1S/C10H14O/c1-10(2,11)8-9-6-4-3-5-7-9/h3-7,11H,8H2,1-2H3

InChI key

RIWRBSMFKVOJMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-1-phenyl-2-propanol was used in the preparation of 2-methyl-1-phenyl-2-propyl bromide[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David C Magri et al.
Organic & biomolecular chemistry, 1(19), 3418-3429 (2003-10-31)
Two dialkyl peroxides, devised as kinetic probes for the heterogeneous electron transfer (ET), are studied using heterogeneous and homogeneous electrochemical techniques. The peroxides react by concerted dissociative ET reduction of the O-O bond. Under heterogeneous conditions, the only products isolated
Takao Raku et al.
Biotechnology letters, 26(8), 665-670 (2004-06-18)
For the purpose of developing a new synthetic polymer containing an asymmetric molecule branch, three racemic alcohols, i.e. 1-phenylethanol, 1-(4-methylphenyl)ethanol and 1-(2-naphthyl)ethanol, were esterified enzymatically with divinyladipate using a lipase from Pseudomonas cepacia. The enzymatic acylation of alcohols produced monoacylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service