Skip to Content
Merck
All Photos(2)

Key Documents

187976

Sigma-Aldrich

3-Phenyl-1-butanol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$380.00

$380.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$380.00

About This Item

Linear Formula:
CH3CH(C6H5)CH2CH2OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$380.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.52 (lit.)

bp

138-140 °C/33 mmHg (lit.)

density

0.972 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

CC(CCO)c1ccccc1

InChI

1S/C10H14O/c1-9(7-8-11)10-5-3-2-4-6-10/h2-6,9,11H,7-8H2,1H3

InChI key

SQGBBDFDRHDJCJ-UHFFFAOYSA-N

General description

3-Phenyl-1-butanol undergoes biotransformation to aldehydes by oxidation in the presence of Gluconobacteroxydans DSM 2343[1]. It undergoes enantioselective transesterification reaction with vinyl acetate catalyzed by lipase isolated from Pseudomonas cepacia[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of vinyl esters on the enantioselectivity of the lipase-catalysed transesterification of alcohols.
Kawasaki M, et al.
Tetrahedron Asymmetry, 12(4), 585-596 (2001)
Chemoselective oxidation of primary alcohols to aldehydes with Gluconobacter oxydans.
Villa R, et al.
Tetrahedron Letters, 43(34), 6059-6061 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service