
p-Toluenesulfonylmethyl isocyanide

Synonym(s):
(p-Tolylsulfonyl)methyl isocyanide, TosMIC, Tosylmethyl isocyanide
Linear Formula:
CH3C6H4SO2CH2NC
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1