Skip to Content
Merck
All Photos(1)

Key Documents

186465

Sigma-Aldrich

N-Cyclohexylhydroxylamine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NHOH · HCl
CAS Number:
Molecular Weight:
151.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

mp

166-171 °C (lit.)

SMILES string

Cl.ONC1CCCCC1

InChI

1S/C6H13NO.ClH/c8-7-6-4-2-1-3-5-6;/h6-8H,1-5H2;1H

InChI key

SSVAHXZUFFSFER-UHFFFAOYSA-N

Application

N-Cyclohexylhydroxylamine hydrochloride was used in the synthesis of:
  • 4-carbethoxy-2-cyclohexyl-5-(2H)-isoxazolone[1]
  • hexahydroisoxazolequinoline analogs[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tricyclic-isoxazolidine analogues via intramolecular 1, 3-dipolar cycloaddition reactions of nitrones.
Saubern S, et al.
Tetrahedron, 66(14), 2761-2767 (2010)
Reactions of 4-carbethoxy-2-cyclohexyl-5 (2H)-isoxazolone.
Wicks JZW and Patel PP.
The Journal of Organic Chemistry, 46(20), 4068-4069 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service