Skip to Content
Merck
All Photos(1)

Key Documents

635936

Sigma-Aldrich

N-Cyclohexylhydroxylamine

Synonym(s):

N-Hydroxycyclohexanamine, N-Hydroxycyclohexylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$162.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$162.00

About This Item

Linear Formula:
C6H11NHOH
CAS Number:
Molecular Weight:
115.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

mp

139-140 °C (lit.)

storage temp.

−20°C

SMILES string

ONC1CCCCC1

InChI

1S/C6H13NO/c8-7-6-4-2-1-3-5-6/h6-8H,1-5H2

InChI key

GUQRKZPMVLRXLT-UHFFFAOYSA-N

Related Categories

General description

N-Cyclohexylhydroxylamine is formed as one of the by-products during the hydrogenation of nitrocyclohexane to the corresponding cyclohexanone oxime catalyzed by palladium/carbon nanotubes.[1]

Application

Reactant for:
  • Aldehyde-catalyzed intermolecular hydroamination
  • Condensation reactions
  • Preparation of cyclin-dependent kinase inhibitors selective for multiple cyclin-dependent kinases for potential use as antitumor agents
  • Preparation of hydroxyurea derivatives as small molecule bradykinin B1 receptor antagonists

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Hydrogenation of nitrocyclohexane to cyclohexanone oxime over Pd/CNT catalyst under mild conditions"
Liao G-H, et al.
Catalysis Communications, 19, 80-84 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service