Skip to Content
Merck
All Photos(3)

Key Documents

182648

Sigma-Aldrich

Poly(vinyl cinnamate)

average Mn 40,000, powder

Synonym(s):

Polyvinyl cinnamate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$364.00
10 G
$481.00

$364.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$364.00
10 G
$481.00

About This Item

Linear Formula:
[CH2CH(O2CCH=CHC6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$364.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

mol wt

average Mn 40,000

SMILES string

C=COC(=O)\C=C\c1ccccc1

InChI

1S/C9H8O2.C2H4O/c10-9(11)7-6-8-4-2-1-3-5-8;1-2-3/h1-7H,(H,10,11);2-3H,1H2/b7-6+;

InChI key

HCCVHWNULIPQNY-UHDJGPCESA-N

General description

Poly (vinyl cinnamate) is a cross-linkable conjugated polymer with a reactive functional group in the main chain. Its backbone contains polyvinyl alcohol and cinnamoyl groups.

Application

Poly (vinyl cinnamate) has been used to study the cyclodimerization of poly (vinyl cinnamate) in solution using pyrylium and thiopyrylium salts. It can also be used to synthesize photocurable polymer gate dielectricsto fabricate cylindrical organic field-effect transistors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–4 of 4 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the glass transition temperature for Product 182648, Poly(vinyl cinnamate)?

    1 answer
    1. Unfortunately, the glass transition temperature is not determined.

      Helpful?

  3. What is the polydispersity of Product 182648, Poly(vinyl cinnamate)?

    1 answer
    1. The polydispersity has been found to be approximately 2.2-2.3.

      Helpful?

  4. What is the solubility of Product 182648, Poly(vinyl cinnamate)?

    1 answer
    1. It is soluble in tetrahydrofuran (THF) and insoluble in water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service