Skip to Content
Merck
All Photos(1)

Key Documents

90502

Sigma-Aldrich

N,N′-Di-sec-butyl-p-phenylenediamine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$50.50
50 ML
$219.00

$50.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
$50.50
50 ML
$219.00

About This Item

Empirical Formula (Hill Notation):
C14H24N2
CAS Number:
Molecular Weight:
220.35
Beilstein:
2805827
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$50.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥94.0% (GC)
95%

form

liquid

refractive index

n20/D 1.539

density

0.942 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CCC(C)Nc1ccc(NC(C)CC)cc1

InChI

1S/C14H24N2/c1-5-11(3)15-13-7-9-14(10-8-13)16-12(4)6-2/h7-12,15-16H,5-6H2,1-4H3

InChI key

FSWDLYNGJBGFJH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service