When a substance is described as "soluble 17.4%" in DMSO, it means that 17.4 grams of the solute can dissolve in 100 milliliters of DMSO. Therefore, in 1 liter (1000 milliliters) of DMSO, approximately 174 grams of the solute can be dissolved.
185809
Cyanuric acid
98%
Synonym(s):
1,3,5-Triazine-2,4,6-triol, 2,4,6-Trihydroxy-1,3,5-triazine
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
Assay
98%
mp
>360 °C (lit.)
solubility
water: soluble 0.2% at 25 °C(lit.)
water: soluble 10% at 150 °C(lit.)
DMSO: soluble 17.4%(lit.)
water: soluble 2.6% at 90 °C(lit.)
DMF: soluble 7.2%(lit.)
acetone: insoluble
alcohol: soluble (hot)(lit.)
benzene: insoluble
chloroform: insoluble
diethyl ether: insoluble
hydrochloric acid: soluble (conc.)
methanol: insoluble (cold)
potassium hydroxide solution: soluble
SMILES string
O=C1NC(=O)NC(=O)N1
InChI
1S/C3H3N3O3/c7-1-4-2(8)6-3(9)5-1/h(H3,4,5,6,7,8,9)
InChI key
ZFSLODLOARCGLH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- iTRAQ-based proteomics analysis reveals the deregulated proteins related to liver toxicity induced by melamine with or without cyanuric acid in mice.: This study used iTRAQ-based proteomics to analyze liver toxicity in mice induced by melamine and cyanuric acid, identifying deregulated proteins associated with liver damage (Yin et al., 2019).
- The contribution of mediated oxidation mechanisms in the electrolytic degradation of cyanuric acid using diamond anodes.: The research investigates the electrolytic degradation of cyanuric acid using diamond anodes, focusing on mediated oxidation mechanisms for effective degradation (Bensalah et al., 2016).
- Encapsulation of [(SO₄)₄(H₂O)₁₁₂]⁸⁻ clusters in a metal organic framework of pyridyl functionalized cyanuric acid based tris-urea.: This paper explores the encapsulation of sulfate clusters in a metal-organic framework using pyridyl-functionalized cyanuric acid, highlighting its potential in material science applications (Dutta et al., 2015).
- Comparing atrazine and cyanuric acid electro-oxidation on mixed oxide and boron-doped diamond electrodes.: This study compares the electro-oxidation of atrazine and cyanuric acid on different electrodes, providing insights into the efficiency and mechanisms of electrochemical degradation processes (Malpass et al., 2013).
- Anion-pi interactions in cyanuric acids: a combined crystallographic and computational study.: The research examines anion-pi interactions in cyanuric acids through crystallographic and computational methods, contributing to the understanding of supramolecular chemistry (Frontera et al., 2005).
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
DMSO: soluble 17.4%(lit.) Does it means 174 g in 1 liter?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
what is the particle size ?
1 answer-
The particle size for this product is not determined.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service