Skip to Content
Merck
All Photos(3)

Key Documents

165816

Sigma-Aldrich

11-Bromoundecanoic acid

technical grade, 95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$73.00

$73.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$73.00

About This Item

Linear Formula:
Br(CH2)10COOH
CAS Number:
Molecular Weight:
265.19
Beilstein:
1767205
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$73.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

95%

bp

173-174 °C/2 mmHg (lit.)

mp

45-48 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

OC(=O)CCCCCCCCCCBr

InChI

1S/C11H21BrO2/c12-10-8-6-4-2-1-3-5-7-9-11(13)14/h1-10H2,(H,13,14)

InChI key

IUDGNRWYNOEIKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

11-Bromoundecanoic acid reacts with potassium salt of dimethyl hydantoin to yield 4,4-dimethyl hydantoin-undecanoic acid[1].

Application

11-Bromoundecanoic acid was used in the synthesis of 11-phenoxyundecyl phosphate[2] and 11-hydroxytetradecanoic acid[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Navarro et al.
Bioorganic & medicinal chemistry, 4(3), 439-443 (1996-03-01)
The synthesis of deuterium labeled 11- and 12-hydroxytetradecanoic acids to study a (11E) desaturase in the moth Spodoptera littoralis is reported. [14,14,14-2H3] 12-hydroxytetradecanoic acid was synthesized in four steps from 11-iodo-1-undecene in 49% overall yield. Deuterium was introduced by reaction
L L Danilov et al.
Bioorganicheskaia khimiia, 35(3), 431-432 (2009-07-22)
A new scheme of synthesis of 11-phenoxyundecyl phosphate from 11-bromoundecanoic acid was suggested for its ability to react as an acceptor of 2-acetamido-2-deoxy-alpha-D-glucopyranosyl phosphate in a reaction catalyzed by UDP-N-acetylglucosamine : polyprenyl phosphate N-acetylglucosamine phosphotransferase from Salmonella arizona O:59.
Revathi V Padmanabhuni et al.
Industrial & engineering chemistry research, 51(14), 5148-5156 (2012-09-04)
The purpose of this study was to demonstrate that the surface of CaCO(3) fillers could be coated with an N-halamine based fatty acid to make the filler surface organophilic and accomplish antibacterial activity simultaneously, rendering the resulting polymer-filler composites antimicrobial.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service