Skip to Content
Merck
All Photos(2)

Key Documents

447463

Sigma-Aldrich

Methyl 11-bromoundecanoate

95%

Synonym(s):

11-Bromoundecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$204.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$204.00

About This Item

Linear Formula:
Br(CH2)10CO2CH3
CAS Number:
Molecular Weight:
279.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$204.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.465 (lit.)

bp

115 °C/0.04 mmHg (lit.)

density

1.157 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

COC(=O)CCCCCCCCCCBr

InChI

1S/C12H23BrO2/c1-15-12(14)10-8-6-4-2-3-5-7-9-11-13/h2-11H2,1H3

InChI key

HFNPVFKUZYCDIB-UHFFFAOYSA-N

Application

Methyl 11-bromoundecanoate can be used as a reactant to synthesize:
  • Methyl 11-(2,5-dibromophenoxy)undecanoate, which is employed as a precursor to prepare acetylenic cyclophanes.[1]
  • Methyl 11-[(1-phenyl-1H-tetrazol-5-yl)thio]undecanoate, a key intermediate applicable in the synthesis of emmyguyacins side chain.[2]
  • Betain derivatives of 11-bromoundecanoic acid, as potential microbial agents.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Three Step Synthesis of 11-Cycloheptylundecanoic Acid, a Component of the Thermoacidophile Alicyclobacillus cycloheptanicus.
Hassarajani SA and Mamdapur VR.
Molecules (Basel), 3(2), 41-43 (1998)
Synthesis, characterization, antimicrobial and anti-biofilm activity of a new class of 11-bromoundecanoic acid-based betaines
Yasa SR, et al.
Medicinal Chemistry Research, 26(10), 2592-2601 (2017)
Collins et al.
Organic letters, 2(20), 3189-3192 (2000-09-29)
The synthesis of a series of novel acetylenic cyclophanes is described. X-ray crystallographic analysis of the core structure revealed a twisted conformation with helical chirality. Preliminary results suggest that these cyclophanes, with appropriate functionality, have the potential to act as
Studies on ω-Oxidation of Fatty Acids in vitro.
KAMEI S, et al.
Journal of Biochemistry, 56(1), 72-76 (1964)
Santanu Jana et al.
Organic letters, 20(21), 6938-6942 (2018-10-24)
Fungal glycolipids emmyguyacins A and B inhibit the pH-dependent conformational change of hemaglutinin A during replication of the Influenza virus. Herein, we report the first total synthesis and structure confirmation of emmyguyacins A and B. Our efficient route, which involves

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service